missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trimethoxybenzaldehyde, 98%

Manufacturer:  Alfa Aesar™ A1123830

 View more versions of this product

Catalog No. AAA1123830


 

Description

Description

Applications
2,4,6-Trimethoxybenzaldehyde was used in the preparation and characterization of three RNA-specific fluorescent probes and their use in live cell imaging. It was used as starting reagent for the regioselective synthesis of new (+/-)-8-alkyl-5,7-dihydroxy-4-(4-hydroxyphenyl)-3,4-dihydrocoumarins.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

2,4,6-Trimethoxybenzaldehyde
250g
98%
MFCD00003313
CRBZVDLXAIFERF-UHFFFAOYSA-N
2,4,6-trimethoxybenzaldehyde
70019
119°C to 123°C
830-79-5
C10H12O4
1956051
COC1=CC(=C(C(=C1)OC)C=O)OC
196.202
196.2

Chemical Identifiers

830-79-5
196.202
CRBZVDLXAIFERF-UHFFFAOYSA-N
C10H12O4
MFCD00003313
70019
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CU8460200

SDS Documents