missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Trimethoxyaniline, 97%, Maybridge

Chemical Identifiers

CAS 14227-17-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H13NO3
Formula Weight 183.21
MDL Number MFCD00017165
Chemical Name or Material 2, 4, 6-Trimethoxyaniline
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
BTB09100DA SDS Fisher Scientific Company
BTB09100DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB09100CB SDS Fisher Scientific Company
BTB09100CB
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

14227-17-9
100.0
C9H13NO3
MFCD00017165
97%
96.5
97%
183.21
2, 4, 6-Trimethoxyaniline

Specifications

1g
96.5
Amber Glass Bottle
C9H13NO3
MFCD00017165
97%
14227-17-9
100.0
97%
183.21
2, 4, 6-Trimethoxyaniline

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning