missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Triisopropylbenzoic acid, 97%

Manufacturer:  Alfa Aesar™ A1548930

 View more versions of this product

Catalog No. AAA1548930



Description

Description

Applications
2,4,6-Triisopropylbenzoic acid is a hindered acid intermediate compound in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

2,4,6-Triisopropylbenzoic acid
250g
C16H24O2
2372331
ULVHAZFBJJXIDO-UHFFFAOYSA-N
2,4,6-tri(propan-2-yl)benzoic acid
96214
97%
184°C to 187°C
49623-71-4
MFCD00015031
2,4,6-triisopropylbenzoic acid, 2,4,6-triisopropyl benzoic acid, 2,4,6-tris 1-methylethyl benzoic acid, 2,4,6-tri propan-2-yl benzoic acid, benzoic acid,2,4,6-tris 1-methylethyl, benzoic acid, 2,4,6-tris 1-methylethyl, 2,4,6-tris methylethyl benzoic acid, acmc-20aoew, pubchem7877
CC(C)C1=CC(=C(C(=C1)C(C)C)C(=O)O)C(C)C
248.366
248.37

Chemical Identifiers

49623-71-4
248.366
ULVHAZFBJJXIDO-UHFFFAOYSA-N
96214
CC(C)C1=CC(=C(C(=C1)C(C)C)C(=O)O)C(C)C
C16H24O2
MFCD00015031
2,4,6-triisopropylbenzoic acid, 2,4,6-triisopropyl benzoic acid, 2,4,6-tris 1-methylethyl benzoic acid, 2,4,6-tri propan-2-yl benzoic acid, benzoic acid,2,4,6-tris 1-methylethyl, benzoic acid, 2,4,6-tris 1-methylethyl, 2,4,6-tris methylethyl benzoic acid, acmc-20aoew, pubchem7877
2,4,6-tri(propan-2-yl)benzoic acid
Documents