missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Triisopropylbenzenesulfonyl Hydrazide, TCI America™

Manufacturer:  TCI America T159125G

 View more versions of this product

Catalog No. T159125G



Specifications

Specifications

2,4,6-Triisopropylbenzenesulfonyl Hydrazide
122°C
39085-59-1
MFCD00014750
2,4,6-triisopropylbenzenesulfonohydrazide, 2,4,6-triisopropylbenzenesulfonyl hydrazide, 2,4,6-triisopropylbenzenesulphonohydrazide, 2,4,6-tri propan-2-yl benzenesulfonohydrazide, benzenesulfonic acid, 2,4,6-tris 1-methylethyl-, hydrazide, tpsh, ksc918s7f, 2,4,6-triisopropylbenzene sulfonohydrazide, 2,4,6-triisopropylbenzenesulfono hydrazide, 2,4,6-triisopropylbenzene sulphonohydrazide
CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)NN)C(C)C
298.445
298.45
≥95.0% (T)
White
25g
C15H26N2O2S
1325
UGRVYFQFDZRNMQ-UHFFFAOYSA-N
2,4,6-tri(propan-2-yl)benzenesulfonohydrazide
359333
Crystal-Powder at 20°C

Chemical Identifiers

39085-59-1
298.445
UGRVYFQFDZRNMQ-UHFFFAOYSA-N
359333
CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)NN)C(C)C
C15H26N2O2S
MFCD00014750
2,4,6-triisopropylbenzenesulfonohydrazide, 2,4,6-triisopropylbenzenesulfonyl hydrazide, 2,4,6-triisopropylbenzenesulphonohydrazide, 2,4,6-tri propan-2-yl benzenesulfonohydrazide, benzenesulfonic acid, 2,4,6-tris 1-methylethyl-, hydrazide, tpsh, ksc918s7f, 2,4,6-triisopropylbenzene sulfonohydrazide, 2,4,6-triisopropylbenzenesulfono hydrazide, 2,4,6-triisopropylbenzene sulphonohydrazide
2,4,6-tri(propan-2-yl)benzenesulfonohydrazide
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

Documents