missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Triisopropylbenzenesulfonyl hydrazide, Alfa Aesar™

As a reagent for generation of diimide

Manufacturer:  Alfa Aesar 1797604

 View more versions of this product

Catalog No. AA1797604



Specifications

Specifications

2,4,6-Triisopropylbenzenesulfonyl hydrazide
110°C to 112°C (decomposition)
39085-59-1
MFCD00014750
2,4,6-triisopropylbenzenesulfonohydrazide, 2,4,6-triisopropylbenzenesulfonyl hydrazide, 2,4,6-triisopropylbenzenesulphonohydrazide, 2,4,6-tri propan-2-yl benzenesulfonohydrazide, benzenesulfonic acid, 2,4,6-tris 1-methylethyl-, hydrazide, tpsh, ksc918s7f, 2,4,6-triisopropylbenzene sulfonohydrazide, 2,4,6-triisopropylbenzenesulfono hydrazide, 2,4,6-triisopropylbenzene sulphonohydrazide
CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)NN)C(C)C
298.445
298.58
Moisture Sensitive
2g
C15H26N2O2S
2145001
UGRVYFQFDZRNMQ-UHFFFAOYSA-N
2,4,6-tri(propan-2-yl)benzenesulfonohydrazide
359333
Crystalline

Chemical Identifiers

39085-59-1
298.445
UGRVYFQFDZRNMQ-UHFFFAOYSA-N
359333
CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)NN)C(C)C
C15H26N2O2S
MFCD00014750
2,4,6-triisopropylbenzenesulfonohydrazide, 2,4,6-triisopropylbenzenesulfonyl hydrazide, 2,4,6-triisopropylbenzenesulphonohydrazide, 2,4,6-tri propan-2-yl benzenesulfonohydrazide, benzenesulfonic acid, 2,4,6-tris 1-methylethyl-, hydrazide, tpsh, ksc918s7f, 2,4,6-triisopropylbenzene sulfonohydrazide, 2,4,6-triisopropylbenzenesulfono hydrazide, 2,4,6-triisopropylbenzene sulphonohydrazide
2,4,6-tri(propan-2-yl)benzenesulfonohydrazide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : Moisture Sensitive

SDS Documents