Learn More
2,4,6-Triisopropylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 152791000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2, 4, 6-Triisopropylbenzenesulfonyl chloride | |
6553-96-4 | |
100.0 | |
White to Beige | |
97% | |
C15H23ClO2S | |
MFCD00007433 | |
01,1228; 03,308; 06,622 | |
Solubility in water: decomposes | |
CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)Cl)C(C)C | |
302.85 | |
302.85 | |
Crystalline Solid |
97% | |
96.0 | |
93°C to 97°C | |
Authentic | |
Glass bottle | |
[(CH3)2CH]3C6H2SO2Cl | |
100 g | |
2,4,6-triisopropylbenzenesulfonyl chloride, 2,4,6-triisopropylbenzene-1-sulfonyl chloride, trisyl chloride, 2,4,6-triisopropylbenzenesulphonyl chloride, benzenesulfonyl chloride, 2,4,6-tris 1-methylethyl, 2,4,6-triisopropylbenzenesulfonylchloride, 2,4,6-triisopropyl-benzenesulfonyl chloride, tpscl 2,4,6-triisopropyl benzenesulfonyl chloride, 2,4,6-triisopropylphenylsulfonyl chloride, 2,4,6-triisopropyl benzenesulfonyl chloride | |
JAPYIBBSTJFDAK-UHFFFAOYSA-N | |
2,4,6-tri(propan-2-yl)benzenesulfonyl chloride | |
81042 | |
97% |
Chemical Identifiers
6553-96-4 | |
302.85 | |
JAPYIBBSTJFDAK-UHFFFAOYSA-N | |
81042 | |
CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)Cl)C(C)C |
C15H23ClO2S | |
MFCD00007433 | |
2,4,6-triisopropylbenzenesulfonyl chloride, 2,4,6-triisopropylbenzene-1-sulfonyl chloride, trisyl chloride, 2,4,6-triisopropylbenzenesulphonyl chloride, benzenesulfonyl chloride, 2,4,6-tris 1-methylethyl, 2,4,6-triisopropylbenzenesulfonylchloride, 2,4,6-triisopropyl-benzenesulfonyl chloride, tpscl 2,4,6-triisopropyl benzenesulfonyl chloride, 2,4,6-triisopropylphenylsulfonyl chloride, 2,4,6-triisopropyl benzenesulfonyl chloride | |
2,4,6-tri(propan-2-yl)benzenesulfonyl chloride |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 229-479-6
TSCA : TSCA
RUO – Research Use Only