Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Triisopropylbenzenesulfonyl azide, 98%, stab. with ca 10% water

$76.39 - $223.00

Chemical Identifiers

Chemical Name or Material 2,4,6-Triisopropylbenzenesulfonyl azide
CAS 36982-84-0
Name Note stabilized with ≈10% water
Molecular Formula C15H23N3O2S
Formula Weight 309.43
MDL Number MFCD00859286
Synonym Trisyl azide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1938306

  SDS
alfa aesar™
L1938306
5g Each for $223.00

AAL1938303

  SDS
alfa aesar™
L1938303
1g Each for $76.39
Description & Specifications

Specifications

Formula Weight 309.43
Chemical Name or Material 2,4,6-Triisopropylbenzenesulfonyl azide
Name Note stabilized with ≈10% water
Assay 98%
Molecular Formula C15H23N3O2S
MDL Number MFCD00859286
CAS 36982-84-0
Synonym Trisyl azide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 40° to 44°C
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C

Applications
2,4,6-Triisopropylbenzenesulfonyl azide is used as a reagent for diazo transfer to ketones, e.g. under phase-transfer conditions, giving better yields of sterically-hindered ɑ-diazo ketones than other methods. It is also used for azide transfer to potassium enolates (KHMDS) to form the ɑ-azido derivatives.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C