Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4',6'-Triisopropylacetophenone, 98+%

$43.41 - $1,870.23

Chemical Identifiers

Chemical Name or Material 2',4',6'-Triisopropylacetophenone
CAS 2234-14-2
Molecular Formula C17H26O
Formula Weight 246.39
MDL Number MFCD00015032
Beilstein 2051003
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1369314

 
alfa aesar™
A1369314
25g Each for $145.92

AAA1369306

 
alfa aesar™
A1369306
5g Each for $43.41

AAA1369322

 
alfa aesar™
A1369322
100g Each for $486.42

AAA1369336

 
alfa aesar™
A1369336
500g Each for $1,870.23
Description & Specifications

Specifications

Formula Weight 246.39
Chemical Name or Material 2',4',6'-Triisopropylacetophenone
Assay ≥98%
Molecular Formula C17H26O
MDL Number MFCD00015032
CAS 2234-14-2
Beilstein 2051003
Melting Point 85° to 88°C
Quantity 25g

Applications
2′,4′,6′-Triisopropylacetophenone is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in refrigerated condition. Incompatible with bases and oxidizing agents.