Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4',6'-Trihydroxyacetophenone, Ultrapure MALDI Matrix

For use as a matrix in MALDI mass spectrometry analysis

Chemical Identifiers

Chemical Name or Material 2',4',6'-Trihydroxyacetophenone
CAS 480-66-0
Name Note Ultrapure MALDI Matrix
Formula Weight 168.15
MDL Number MFCD00149091
Beilstein 1911197
Synonym THAP
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67719TDL

 
alfa aesar™
J67719TDL
90mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 168.15
Melting Point 218° to 223°C
Chemical Name or Material 2',4',6'-Trihydroxyacetophenone
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
TSCA TSCA
Physical Form Powder
MDL Number MFCD00149091
CAS 480-66-0
Synonym THAP
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Color Pale yellow
Name Note Ultrapure MALDI Matrix
Beilstein 1911197
RTECS Number AN0528000
Quantity 90mg
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.