Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trifluoropyridine, 97%

Chemical Identifiers

Chemical Name or Material 2,4,6-Trifluoropyridine
CAS 3512-17-2
Molecular Formula C5H2F3N
Formula Weight 133.07
MDL Number MFCD03001161
UN Number UN2924
Beilstein 1364338
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1954103

  SDS
alfa aesar™
L1954103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 133.07
MDL Number MFCD03001161
Beilstein 1364338
Chemical Name or Material 2,4,6-Trifluoropyridine
Molecular Formula C5H2F3N
CAS 3512-17-2
Assay 97%
UN Number UN2924
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Flash Point 17°C (62°F)
Boiling Point 102°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.