Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trifluorophenylhydrazine, 97%

$79.58 - $190.86

Chemical Identifiers

Chemical Name or Material 2,4,6-Trifluorophenylhydrazine
CAS 80025-72-5
Molecular Formula C6H5F3N2
Formula Weight 162.11
MDL Number MFCD03095329
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2610403

 
alfa aesar™
H2610403
1g Each for $190.86

AAH26104MD

 
alfa aesar™
H26104MD
250mg Each for $79.58
Description & Specifications

Specifications

Formula Weight 162.11
Chemical Name or Material 2,4,6-Trifluorophenylhydrazine
Assay 97%
Molecular Formula C6H5F3N2
MDL Number MFCD03095329
CAS 80025-72-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 97° to 101°C
Quantity 1g

Applications
Propionic acid (2,4,6-trichlorophenyl)hydrazide was prepared by reaction of (2,4,6-trichlorophenyl)- hydrazine with propionyl chloride in pyridine. Visnaginone Ia reacted with phenyl hydrazine and 2,4,6- trichlorophenyl hydrazine to yield the corresponding hydrazones.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.