Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trifluorophenylacetonitrile, 98%

Chemical Identifiers

Chemical Name or Material 2,4,6-Trifluorophenylacetonitrile
CAS 220227-80-5
Molecular Formula C8H4F3N
Formula Weight 171.12
MDL Number MFCD00061221
UN Number UN3276
Synonym 2,4,6-Trifluorobenzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2017102

  SDS
alfa aesar™
B2017102
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2017188

  SDS
alfa aesar™
B2017188
2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.12
Chemical Name or Material 2,4,6-Trifluorophenylacetonitrile
Assay 98%
Molecular Formula C8H4F3N
MDL Number MFCD00061221
CAS 220227-80-5
UN Number UN3276
Synonym 2,4,6-Trifluorobenzyl cyanide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Quantity 0.5g

Applications
2,4,6-Trifluorophenylacetonitrile is used in agrochemical, pharmaceutical and dyestuff field

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.