Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trifluorophenylacetic acid, 98%

$107.30 - $427.92

Chemical Identifiers

Chemical Name or Material 2,4,6-Trifluorophenylacetic acid
CAS 209991-63-9
Molecular Formula C8H5F3O2
Formula Weight 190.12
MDL Number MFCD00061218
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2003902

 
alfa aesar™
B2003902
0.5g Each for $107.30

AAB2003988

 
alfa aesar™
B2003988
2.5g Each for $427.92
Description & Specifications

Specifications

Formula Weight 190.12
Chemical Name or Material 2,4,6-Trifluorophenylacetic acid
Assay 98%
Molecular Formula C8H5F3O2
MDL Number MFCD00061218
CAS 209991-63-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 110° to 112°C
Quantity 0.5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.