Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trifluorophenyl isocyanate, tech. 85%

Chemical Identifiers

Chemical Name or Material 2,4,6-Trifluorophenyl isocyanate
CAS 50528-80-8
Purity Grade Technical Grade
Molecular Formula C7H2F3NO
Formula Weight 173.11
MDL Number MFCD03426028
UN Number UN3080
Beilstein 6325663
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1934903

  SDS
alfa aesar™
L1934903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1934906

  SDS
alfa aesar™
L1934906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 173.11
Chemical Name or Material 2,4,6-Trifluorophenyl isocyanate
Assay 85%
Molecular Formula C7H2F3NO
MDL Number MFCD03426028
Health Hazard 2 GHS H Statement
H330-H334-H340-H350-H372-H226
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P304+P340-P320-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Purity Grade Technical Grade
CAS 50528-80-8
UN Number UN3080
Beilstein 6325663
Health Hazard 1 Danger
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, FLAMMABLE, N.O.S.
Quantity 1g

Applications
2,4,6-Trifluorophenyl isocyanate is used as a pharmaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, amines and alcohols. It is sensitive to moisture. Store under dry inert gas.

Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P304+P340-P320-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H330-H334-H340-H350-H372-H226
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Flammable liquid and vapor.

2° to 8°C