missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trifluorophenol, 99%

Manufacturer:  Alfa Aesar™ L1128503

 View more versions of this product

Catalog No. AAL1128503



Specifications

Specifications

2,4,6-Trifluorophenol
1g
C6H3F3O
UN3261
phenol, 2,4,6-trifluoro, 2,4,6-trifluoro-phenol, pubchem2317, acmc-1cod0, phenol,2,4,6-trifluoro, 2,4,6-trifluorophenol
C1=C(C=C(C(=C1F)O)F)F
148.084
148.09
47°C to 52°C
2268-17-9
MFCD00061216
2089551
QQFWMPUXPLBWTG-UHFFFAOYSA-N
2,4,6-trifluorophenol
519974
99%

Chemical Identifiers

2268-17-9
148.084
QQFWMPUXPLBWTG-UHFFFAOYSA-N
519974
C1=C(C=C(C(=C1F)O)F)F
C6H3F3O
MFCD00061216
phenol, 2,4,6-trifluoro, 2,4,6-trifluoro-phenol, pubchem2317, acmc-1cod0, phenol,2,4,6-trifluoro, 2,4,6-trifluorophenol
2,4,6-trifluorophenol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Documents