Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trifluorobenzotrifluoride, 99%

Chemical Identifiers

Chemical Name or Material 2,4,6-Trifluorobenzotrifluoride
CAS 122030-04-0
Molecular Formula C7H2F6
Formula Weight 200.08
MDL Number MFCD00190109
UN Number UN1993
Synonym alpha,alpha,alpha,2,4,6-Hexafluorotoluene; 1,3,5-Trifluoro-2-(trifluoromethyl)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1816703

  SDS
alfa aesar™
L1816703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1816706

  SDS
alfa aesar™
L1816706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1816714

  SDS
alfa aesar™
L1816714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.08
CAS 122030-04-0
Assay 99%
Molecular Formula C7H2F6
Chemical Name or Material 2,4,6-Trifluorobenzotrifluoride
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym alpha,alpha,alpha,2,4,6-Hexafluorotoluene; 1,3,5-Trifluoro-2-(trifluoromethyl)benzene
MDL Number MFCD00190109
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Flash Point 22°C (71°F)
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Health Hazard 1 Danger
Boiling Point 105° to 107°C
UN Number UN1993

Applications
2,4,6-Trifluorobenzotrifluoride finds its application as an intermediate in organic syntheses.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges.

Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.