Please login to your online account to display your discounted pricing

2,4,6-Trifluorobenzaldehyde 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4, 6-Trifluorobenzaldehyde
CAS 58551-83-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C7H3F3O
Formula Weight 160.09
MDL Number MFCD00061197
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC384200050

  SDS
thermo fisher scientific
AC384200050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC384200010

  SDS
thermo fisher scientific
AC384200010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0% (GC)
Color White to slightly grey
Physical Form Powder
Formula Weight 160.09
Quantity 5g
Melting Point 62° to 67°C
CAS Max % 100.0
CAS Min % 96.0
CAS 58551-83-0
Chemical Name or Material 2, 4, 6-Trifluorobenzaldehyde
Infrared Spectrum Authentic
Packaging Glass bottle
MDL Number MFCD00061197
Molecular Formula C7H3F3O
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.