Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trifluorobenzaldehyde, 97%

$96.51 - $871.23

Chemical Identifiers

Chemical Name or Material 2,4,6-Trifluorobenzaldehyde
CAS 58551-83-0
Molecular Formula C7H3F3O
Formula Weight 160.1
MDL Number MFCD00061197
Beilstein 6800184
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2011503

 
alfa aesar™
B2011503
1g Each for $96.51

AAB2011506

 
alfa aesar™
B2011506
5g Each for $374.04

AAB2011514

 
alfa aesar™
B2011514
25g Each for $871.23
Description & Specifications

Specifications

Formula Weight 160.1
Chemical Name or Material 2,4,6-Trifluorobenzaldehyde
Assay 97%
Molecular Formula C7H3F3O
MDL Number MFCD00061197
CAS 58551-83-0
Beilstein 6800184
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Boiling Point 165°C
Melting Point 63° to 68°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.