missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4',6'-Trifluoroacetophenone, 98%

$62.22 - $184.00

Chemical Identifiers

CAS 51788-77-3
Molecular Formula C8H5F3O
Formula Weight 174.12
MDL Number MFCD00061194
Chemical Name or Material 2',4',6'-Trifluoroacetophenone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2619806 SDS Alfa Aesar™
H2619806
5g Each for $184.00
AAH2619803 SDS Alfa Aesar™
H2619803
1g Each for $62.22
Specifications

Description

Applications
2′,4′,6′-Trifluoroacetophenone was the reactant in the synthesis of 4, 6-and 6, 7-difluoro-3-methyl-1H-indazoles.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

51788-77-3
174.12
2',4',6'-Trifluoroacetophenone
C8H5F3O
MFCD00061194

Specifications

5g
C8H5F3O
MFCD00061194
98%
51788-77-3
174.12
2',4',6'-Trifluoroacetophenone

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning