Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trichloropyrimidine-5-carboxaldehyde, 97%

Chemical Identifiers

Chemical Name or Material 2,4,6-Trichloropyrimidine-5-carboxaldehyde
CAS 50270-27-4
Formula Weight 211.43
MDL Number MFCD02257700
UN Number UN2811
Beilstein 744505
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6486603

  SDS
alfa aesar™
H6486603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6486606

  SDS
alfa aesar™
H6486606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 211.43
Melting Point 131° to 132°C
UN Number UN2811
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Health Hazard 1 Warning
Chemical Name or Material 2,4,6-Trichloropyrimidine-5-carboxaldehyde
MDL Number MFCD02257700
CAS 50270-27-4
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Storage Note 1 -30° to -10°C
Beilstein 744505
Assay 97%
Color White to brown
Physical Form Solid

Applications
It is used as pharmaceutical intermediate. It is a versatile intermediate in the synthesis of reactive dyes.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store in cool, well ventilated area. Store cold 2-4°C.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

-30° to -10°C