Please login to your online account to display your discounted pricing

2,4,6-Trichlorophenylhydrazine, 98%, ACROS Organics™


Quantity 5g

Manufacturer: fisher scientific acros  421580050

 View more versions of this product

Catalog No. AC421580050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Melting Point 140° to 143°C
Color Beige
Quantity 5g
Chemical Name or Material 2, 4, 6-Trichlorophenylhydrazine
Name Note 98%
CAS 1252756
CAS Min % 97.5
CAS Max % 100.0
Physical Form Beige powder
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C6H5Cl3N2
Formula Weight 211.48g/mol
MDL Number MFCD00025090
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 226-217-2
TSCA TSCA

Specifications

Chemical Name or Material 2, 4, 6-Trichlorophenylhydrazine
Name Note 98%
CAS 1252756
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C6H5Cl3N2
Formula Weight 211.48g/mol
MDL Number MFCD00025090
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

SDS & Safety Documents