missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trichlorophenyl isothiocyanate, 98%

Manufacturer:  Alfa Aesar™ L1211806

 View more versions of this product

Catalog No. AAL1211806



Specifications

Specifications

2,4,6-Trichlorophenyl isothiocyanate
72°C to 76°C
22134-07-2
MFCD00041053
880716
DSXVZIOQROMOEF-UHFFFAOYSA-N
1,3,5-trichloro-2-isothiocyanatobenzene
123399
98%
Irritating
5g
C7H2Cl3NS
UN2923
2,4,6-trichlorophenyl isothiocyanate, benzene,1,3,5-trichloro-2-isothiocyanato, benzene, 1,3,5-trichloro-2-isothiocyanato, 2,4,6-trichlorophenyl isothiocyanate, 2,4,6-trichlorophenylisothiocyanate, acmc-1cngp, 2,4,6-trichlorobenzenisothiocyanate, #, 2,4,6-trichlorophenyl isothiocyanate
C1=C(C=C(C(=C1Cl)N=C=S)Cl)Cl
238.51
238.52

Chemical Identifiers

22134-07-2
238.51
DSXVZIOQROMOEF-UHFFFAOYSA-N
123399
C1=C(C=C(C(=C1Cl)N=C=S)Cl)Cl
C7H2Cl3NS
MFCD00041053
2,4,6-trichlorophenyl isothiocyanate, benzene,1,3,5-trichloro-2-isothiocyanato, benzene, 1,3,5-trichloro-2-isothiocyanato, 2,4,6-trichlorophenyl isothiocyanate, 2,4,6-trichlorophenylisothiocyanate, acmc-1cngp, 2,4,6-trichlorobenzenisothiocyanate, #, 2,4,6-trichlorophenyl isothiocyanate
1,3,5-trichloro-2-isothiocyanatobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

Documents