missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Trichlorophenyl Isocyanate, TCI America™

$114.58 - $381.95

Chemical Identifiers

CAS 2505-31-9
Molecular Formula C7H2Cl3NO
Molecular Weight (g/mol) 222.449
MDL Number MFCD00013845
InChI Key VDYWXVDWKFAUKE-UHFFFAOYSA-N
Synonym Isocyanic Acid 2,4,6-Trichlorophenyl Ester
PubChem CID 613574
IUPAC Name 1,3,5-trichloro-2-isocyanatobenzene
SMILES C1=C(C=C(C(=C1Cl)N=C=O)Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I05655G View Documents TCI America
I05655G
5g Each for $381.95
 
I05651G View Documents TCI America
I05651G
1g Each for $114.58
 
Specifications

Chemical Identifiers

2505-31-9
222.449
VDYWXVDWKFAUKE-UHFFFAOYSA-N
613574
C1=C(C=C(C(=C1Cl)N=C=O)Cl)Cl
C7H2Cl3NO
MFCD00013845
Isocyanic Acid 2,4,6-Trichlorophenyl Ester
1,3,5-trichloro-2-isocyanatobenzene

Specifications

101°C
67°C
2505-31-9
MFCD00013845
Isocyanic Acid 2,4,6-Trichlorophenyl Ester
C1=C(C=C(C(=C1Cl)N=C=O)Cl)Cl
222.449
222.45
≥98.0% (GC)
Undesignated
5g
C7H2Cl3NO
2206
VDYWXVDWKFAUKE-UHFFFAOYSA-N
1,3,5-trichloro-2-isocyanatobenzene
613574
Crystal-Powder at 20°C
2,4,6-Trichlorophenyl Isocyanate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator