Please login to your online account to display your discounted pricing

2,4,6-Trichlorobenzoyl chloride, 98%, ACROS Organics™

$72.15 - $72.15

Chemical Identifiers

Chemical Name or Material 2, 4, 6-Trichlorobenzoyl chloride
CAS 4136-95-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H2Cl4O
Formula Weight 243.9
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC335180050

  SDS
thermo fisher scientific
33518-0050
Glass bottle 5g Each for $72.15

AC335180250

  SDS
thermo fisher scientific
AC335180250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 243.9
Physical Form Liquid
Quantity 5g
Color Clear yellow
Density 1.56g/mL
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.5750 to 1.5770 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 1 Danger
CAS 4136-95-2
Assay Percent Range 98%
Chemical Name or Material 2, 4, 6-Trichlorobenzoyl chloride
Molecular Formula C7H2Cl4O
CAS Min % 97.5
Infrared Spectrum Authentic
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.