Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trichlorobenzoic acid, 94%

$65.09 - $65.09

Chemical Identifiers

Chemical Name or Material 2,4,6-Trichlorobenzoic acid
CAS 50-43-1
Molecular Formula C7H3Cl3O2
Formula Weight 225.46
MDL Number MFCD00060699
Beilstein 1874127
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1419106

  SDS
alfa aesar™
L1419106
5g Each for $65.09
Description & Specifications

Specifications

Formula Weight 225.46
Chemical Name or Material 2,4,6-Trichlorobenzoic acid
CAS 50-43-1
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 94%
Health Hazard 1 Warning
Molecular Formula C7H3Cl3O2
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number DH8050000
Melting Point 162° to 164°C
Beilstein 1874127
MDL Number MFCD00060699
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.