missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trichlorobenzenesulfonamide, 97%

$132.16 - $478.05

Chemical Identifiers

CAS 28460-30-2
Molecular Formula C6H4Cl3NO2S
Molecular Weight (g/mol) 260.513
MDL Number MFCD00955746
InChI Key IPAFNZVMBOZODU-UHFFFAOYSA-N
Synonym 2,4,6-trichlorobenzene-1-sulfonamide
PubChem CID 1268283
IUPAC Name 2,4,6-trichlorobenzenesulfonamide
SMILES C1=C(C=C(C(=C1Cl)S(=O)(=O)N)Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2693403 View Documents Alfa Aesar™
H2693403
1g Each for $132.16
 
AAH2693406 View Documents Alfa Aesar™
H2693406
5g Each for $478.05
 
Specifications

Chemical Identifiers

28460-30-2
260.513
IPAFNZVMBOZODU-UHFFFAOYSA-N
1268283
C1=C(C=C(C(=C1Cl)S(=O)(=O)N)Cl)Cl
C6H4Cl3NO2S
MFCD00955746
2,4,6-trichlorobenzene-1-sulfonamide
2,4,6-trichlorobenzenesulfonamide

Specifications

179°C to 181°C
28460-30-2
MFCD00955746
IPAFNZVMBOZODU-UHFFFAOYSA-N
2,4,6-trichlorobenzenesulfonamide
1268283
97%
1g
C6H4Cl3NO2S
2,4,6-trichlorobenzene-1-sulfonamide
C1=C(C=C(C(=C1Cl)S(=O)(=O)N)Cl)Cl
260.513
260.53
2,4,6-Trichlorobenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning