missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Trichloro-5-pyrimidinecarboxaldehyde, TCI America™

Manufacturer:  TCI America T31005G

 View more versions of this product

Catalog No. T31005G



Specifications

Specifications

2,4,6-Trichloro-5-pyrimidinecarboxaldehyde
132°C
50270-27-4
MFCD02257700
KVJIRFGNHAAUNQ-UHFFFAOYSA-N
2,4,6-trichloropyrimidine-5-carbaldehyde
10932746
Crystal-Powder at 20°C
White-Yellow
5g
C5HCl3N2O
2,4,6-trichloro-5-pyrimidinecarboxaldehyde, 2,4,6-trichloro-pyrimidine-5-carbaldehyde, 2,4,6-trichloro-5-formylpyrimidine, 5-formyl-2,4,6-trichloropyrimidine, 2,4,6-trichloro-pyrimidine-5-carbal, 2,4,6-trichloropyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 2,4,6-trichloro, 5-pyrimidinecarboxaldehyde,2,4,6-trichloro, pubchem13189, 2,4,6-tri-chloropyrimidine-5-carbaldehyde
C(=O)C1=C(N=C(N=C1Cl)Cl)Cl
211.426
211.43
≥98.0% (GC)

Chemical Identifiers

50270-27-4
211.426
KVJIRFGNHAAUNQ-UHFFFAOYSA-N
10932746
C(=O)C1=C(N=C(N=C1Cl)Cl)Cl
C5HCl3N2O
MFCD02257700
2,4,6-trichloro-5-pyrimidinecarboxaldehyde, 2,4,6-trichloro-pyrimidine-5-carbaldehyde, 2,4,6-trichloro-5-formylpyrimidine, 5-formyl-2,4,6-trichloropyrimidine, 2,4,6-trichloro-pyrimidine-5-carbal, 2,4,6-trichloropyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 2,4,6-trichloro, 5-pyrimidinecarboxaldehyde,2,4,6-trichloro, pubchem13189, 2,4,6-tri-chloropyrimidine-5-carbaldehyde
2,4,6-trichloropyrimidine-5-carbaldehyde
Safety and Handling

Safety and Handling

TSCA : No

Documents