Please login to your online account to display your discounted pricing

2,4,6-Tribromotoluene 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4, 6-Tribromotoluene
CAS 6320-40-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H5Br3
Linear Formula CH3C6H2Br3
Formula Weight 328.83
MDL Number MFCD00013527
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC393380250

  SDS
thermo fisher scientific
AC393380250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC393380050

  SDS
thermo fisher scientific
AC393380050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder and/or Chunks
Formula Weight 328.83
Quantity 25g
Color Yellow to light brown
Melting Point 64° to 70°C
CAS Max % 100.0
CAS Min % 96.0
CAS 6320-40-7
Chemical Name or Material 2, 4, 6-Tribromotoluene
MDL Number MFCD00013527
Packaging Glass bottle
Linear Formula CH3C6H2Br3
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C7H5Br3
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.