Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Tribromopyrimidine, 95%

$238.26 - $238.26

Chemical Identifiers

Chemical Name or Material 2,4,6-Tribromopyrimidine
CAS 36847-11-7
Molecular Formula C4HBr3N2
Formula Weight 316.78g/mol
MDL Number MFCD00233942
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27471MD

 
alfa aesar™
H27471-MD
250mg Each for $238.26
Description & Specifications

Specifications

Quantity 250mg
Chemical Name or Material 2,4,6-Tribromopyrimidine
CAS 36847-11-7
Assay 95%
Molecular Formula C4HBr3N2
Formula Weight 316.78g/mol
MDL Number MFCD00233942
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.