missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Tribromophenyl Acrylate, TCI America™

Manufacturer:  TCI America T313025G

 View more versions of this product

Catalog No. T313025G



Specifications

Specifications

2,4,6-Tribromophenyl Acrylate
78°C
3741-77-3
MFCD00078430
CNLVUQQHXLTOTC-UHFFFAOYSA-N
(2,4,6-tribromophenyl) prop-2-enoate
77343
Crystal-Powder at 20°C
White
25g
C9H5Br3O2
Acrylic Acid 2,4,6-Tribromophenyl Ester
C=CC(=O)OC1=C(C=C(C=C1Br)Br)Br
384.849
384.85
≥98.0% (GC)

Chemical Identifiers

3741-77-3
384.849
CNLVUQQHXLTOTC-UHFFFAOYSA-N
77343
C=CC(=O)OC1=C(C=C(C=C1Br)Br)Br
C9H5Br3O2
MFCD00078430
Acrylic Acid 2,4,6-Tribromophenyl Ester
(2,4,6-tribromophenyl) prop-2-enoate
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0972&(3)-2988&(3)-0961

TSCA : Yes

Documents