missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Tribromophenol, 98%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 160105000

 View more versions of this product

Catalog No. AC160105000



Specifications

Specifications

2, 4, 6-Tribromophenol
Plastic bottle
244.0°C
89.0°C to 93.0°C
98%
97.5
98%
Br3C6H2OH
06,203
15,9775
Solubility in water: 71mg/l (15°C). Other solubilities: soluble in acetone,diethylether,petroleum ether,,benzene,toluene,alcohols,chloroform,pyridine,and carbon tetrachloride
C1=C(C=C(C(=C1Br)O)Br)Br
330.79
CHEBI:47696
Powder
Authentic
244°C
White to Brown
500g
118-79-6
100.0
C6H3Br3O
MFCD00002150
02,424
5175-83-7 bismuth 3+ salt
BSWWXRFVMJHFBN-UHFFFAOYSA-N
2,4,6-tribromophenol
1483
330.79
98%

Chemical Identifiers

118-79-6
330.79
BSWWXRFVMJHFBN-UHFFFAOYSA-N
1483
2,4,6-tribromophenol
C6H3Br3O
MFCD00002150
5175-83-7 bismuth 3+ salt
CHEBI:47696
C1=C(C=C(C(=C1Br)O)Br)Br
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Causes serious eye irritation.
Very toxic to aquatic life.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/ey

GHS Signal Word: Warning

EINECSNumber : 204-278-6

RTECSNumber : SN1225000

TSCA : TSCA

SDS Documents