Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Tribromoanisole, 99%

Chemical Identifiers

Chemical Name or Material 2,4,6-Tribromoanisole
CAS 607-99-8
Formula Weight 344.83
MDL Number MFCD00192510
Beilstein 2210361
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5568403

 
alfa aesar™
H5568403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5568406

 
alfa aesar™
H5568406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5568414

 
alfa aesar™
H5568414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 344.83
Melting Point 84° to 88°C
Boiling Point 297° to 299°C
Chemical Name or Material 2,4,6-Tribromoanisole
Physical Form Solid
Assay 99%
MDL Number MFCD00192510
Beilstein 2210361
Color White to pale brown
Quantity 1g
TSCA TSCA
CAS 607-99-8