missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Tribromoanisole, 99%

Manufacturer:  Alfa Aesar™ H5568403

 View more versions of this product

Catalog No. AAH5568403



Specifications

Specifications

2,4,6-Tribromoanisole
Brown to White
1g
C7H5Br3O
2210361
YXTRCOAFNXQTKL-UHFFFAOYSA-N
1,3,5-tribromo-2-methoxybenzene
11839
Solid
297°C to 299°C
84°C to 88°C
607-99-8
MFCD00192510
1,3,5-tribromo-2-methoxy-benzene
COC1=C(C=C(C=C1Br)Br)Br
344.828
344.83
99%

Chemical Identifiers

607-99-8
344.828
YXTRCOAFNXQTKL-UHFFFAOYSA-N
11839
COC1=C(C=C(C=C1Br)Br)Br
C7H5Br3O
MFCD00192510
1,3,5-tribromo-2-methoxy-benzene
1,3,5-tribromo-2-methoxybenzene
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents