missing translation for 'onlineSavingsMsg'
Learn More
missing translation for 'pleaseLogIn'

Alfa Aesar™ 2,4,6-Tribromo-3-methylphenol, 98+%

$78.04 - $213.96

Chemical Identifiers

CAS 4619-74-3
Molecular Formula C7H5Br3O
Formula Weight 344.84
MDL Number MFCD00019976
Beilstein 1949974
Merck Index 14,9612
Synonym 2,4,6-Tribromo-m-cresol
Chemical Name or Material 2,4,6-Tribromo-3-methylphenol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAL0918314 SDS Alfa Aesar™
L0918314
25g Each for $78.04
AAL0918322 SDS Alfa Aesar™
L0918322
100g Each for $213.96
Specifications Description & Specifications

Chemical Identifiers

4619-74-3
344.84
1949974
2,4,6-Tribromo-m-cresol
C7H5Br3O
MFCD00019976
14,9612
2,4,6-Tribromo-3-methylphenol

Specifications

4619-74-3
344.84
1949974
2,4,6-Tribromo-m-cresol
25g
≥98%
C7H5Br3O
MFCD00019976
14,9612
81° to 83°C
2,4,6-Tribromo-3-methylphenol

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : GP3500000