Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Tribromo-3-methylaniline, 98+%

$39.88 - $321.72

Chemical Identifiers

Chemical Name or Material 2,4,6-Tribromo-3-methylaniline
CAS 71642-16-5
Molecular Formula C7H6Br3N
Formula Weight 343.84
MDL Number MFCD00014768
Beilstein 3252610
Synonym 3-Methyl-2,4,6-tribromoaniline; 2,4,6-Tribromo-m-toluidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1250106

 
alfa aesar™
A1250106
5g Each for $39.88

AAA1250122

 
alfa aesar™
A1250122
100g Each for $321.72
Description & Specifications

Specifications

Formula Weight 343.84
Chemical Name or Material 2,4,6-Tribromo-3-methylaniline
Assay ≥98%
Molecular Formula C7H6Br3N
MDL Number MFCD00014768
CAS 71642-16-5
Beilstein 3252610
Synonym 3-Methyl-2,4,6-tribromoaniline; 2,4,6-Tribromo-m-toluidine
Health Hazard 1 Warning
Melting Point 99° to 102°C
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.