Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Triaminopyrimidine, 98+%

$50.03 - $50.03

Chemical Identifiers

Chemical Name or Material 2,4,6-Triaminopyrimidine
CAS 1004-38-2
Molecular Formula C4H7N5
Formula Weight 125.14
MDL Number MFCD00006100
Beilstein 118448
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2336009

 
alfa aesar™
B2336009
10g Each for $50.03
Description & Specifications

Specifications

Formula Weight 125.14
Chemical Name or Material 2,4,6-Triaminopyrimidine
MDL Number MFCD00006100
Beilstein 118448
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay ≥98%
Molecular Formula C4H7N5
CAS 1004-38-2
Melting Point 248° to 252°C
Quantity 10g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.