missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Triaminopyrimidine, 97%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 139251000

 View more versions of this product

Catalog No. AC139251000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
125.13
Off-white to beige
100.0
Plastic bottle
97%
97%
C4H7N5
Powder
100g
246° to 252°C
96.0
Authentic
2, 4, 6-Triaminopyrimidine
1004-38-2
MFCD00006100

Chemical Identifiers

1004-38-2
100.0
C4H7N5
MFCD00006100
≥96.0%
96.0
97%
125.13
2, 4, 6-Triaminopyrimidine, 97%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 213-720-7

TSCA : TSCA