Please login to your online account to display your discounted pricing

2,4,6-Tri-tert-butylphenol, 97%, ACROS Organics™

$45.38 - $45.38

Chemical Identifiers

Chemical Name or Material 2, 4, 6-Tri-tert-butylphenol
CAS 732-26-3
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C18H30O
Linear Formula [(CH3)3C]3C6H2OH
Formula Weight 262.42
MDL Number MFCD00008821
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC139335000

  SDS
thermo fisher scientific
AC139335000
Plastic bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC139330050

  SDS
thermo fisher scientific
139330050
Glass Bottle 5g Each for $45.38

AC139330500

  SDS
thermo fisher scientific
139330500
Plastic bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500g
Formula Weight 262.42
Beilstein 06, IV, 3539
CAS Min % 96.0
CAS Max % 100.0
UN Number 3077
Packaging Plastic bottle
Linear Formula [(CH3)3C]3C6H2OH
Assay Percent Range 97%
Molecular Formula C18H30O
CAS 732-26-3
Chemical Name or Material 2, 4, 6-Tri-tert-butylphenol
Health Hazard 1 Warning
MDL Number MFCD00008821
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Causes skin irritation.