Please login to your online account to display your discounted pricing

2,4,6-Tri-tert-butylnitrosobenzene, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4, 6-Tri-tert-butylnitrosobenzene
Name Note 99%
CAS 24973-59-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C18H29NO
Linear Formula {(CH3)3C}3C6H2NO
Formula Weight 275.42
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC207980010

  SDS
thermo fisher scientific
207980010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC207982500

  SDS
thermo fisher scientific
207982500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 275.42
Physical Form Powder
Quantity 1g
Color Green
TSCA TSCA
EINECS Number 246-553-3
Melting Point 170° to 173°C
CAS Max % 100.0
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym BNB
Molecular Formula C18H29NO
Chemical Name or Material 2, 4, 6-Tri-tert-butylnitrosobenzene
MDL Number MFCD00008817
Linear Formula {(CH3)3C}3C6H2NO
CAS 24973-59-9
Assay Percent Range 99%
Name Note 99%