Please login to your online account to display your discounted pricing

2,4,6-Collidine, 99%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material 2,4,6-Collidine
CAS 108-75-8
Molecular Formula C8H11N
Formula Weight 121.18
MDL Number MFCD00006338
UN Number UN1992
Beilstein 107283
Merck Index 14,9718
Synonym 2,4,6-Trimethylpyridine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA11058AE

  SDS
alfa aesar
A11058AE
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 121.18
Quantity 100mL
MDL Number MFCD00006338
CAS 108-75-8
Health Hazard 2 GHS H Statement
H311-H226-H302-H315-H319-H335;
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point −43°C
Merck Index 14,9718
Beilstein 107283
Assay 99%
Odor Unpleasant
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
RTECS Number UU0970000
UN Number UN1992
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
TSCA TSCA
Synonym 2,4,6-Trimethylpyridine
Boiling Point 171° to 172°C
pH 9 to 10
Chemical Name or Material 2,4,6-Collidine
Flash Point 54°C (129°F)
Molecular Formula C8H11N

Applications
2,4,6-Collidine is used as a tissue fixative for electron microscopy. It is useful in dehydrohalogenation reactions and acts as a solvent for the cleavage of hindered esters by anhydrous lithium iodide.

Solubility
Slightly miscible with water. Miscible with ether, methanol, acetone, chloroform, benzene, toluene and dilute acids.

Notes
Incompatible with oxidizing agents and strong acids.

Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H311-H226-H302-H315-H319-H335;
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.