missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Collidine, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 161955000

 View more versions of this product

Catalog No. AC161955000


 

Specifications Description & Specifications

Specifications

Liquid
≥98.5%
Clear colorless to yellow
15, 9890
100.0
2, 4, 6-Collidine
C8H11N
108-75-8
98.5
Authentic
1.4970 to 1.4990 (20°C, 589nm)
121.18
500mL
20, 250
01,155; 13,302
2, 4, 6-Trimethylpyridine
(C5H2N)(CH3)3
99%
MFCD00006338
This product darkens on standing
Glass bottle

Chemical Identifiers

108-75-8
100.0
C8H11N
121.18
2, 4, 6-Trimethylpyridine
≥98.5%
98.5
99%
(C5H2N)(CH3)3
MFCD00006338
2, 4, 6-Collidine

Safety and Handling

SDS & Safety Documents

GHS P Statement
Flammable liquid and vapour.
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 203-613-3

RTECSNumber : UU0970000

TSCA : TSCA