missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Collidine, 99%, ACROS Organics™

Manufacturer:  Acros Organics 161951000

 View more versions of this product

Catalog No. AC161951000



Specifications

Specifications

2, 4, 6-Collidine
57°C
Glass bottle
0.91
Colorless to Yellow
100mL
98.5
99%
(C5H2N)(CH3)3
20, 250
15, 9890
BWZVCCNYKMEVEX-UHFFFAOYSA-N
2,4,6-trimethylpyridine
7953
Liquid
0.9100g/mL
Authentic
1.4995 to 1.5015
171.0°C to 172.0°C
-43.0°C
108-75-8
100.0
C8H11N
MFCD00006338
01,155; 13,302
2,4,6-collidine, gamma-collidine, s-collidine, pyridine, 2,4,6-trimethyl, collidine, sym-collidine, 2,4,6-kollidin, unii-7ie4bk5j5v, 2,4,6-trimethyl-pyridine, .gamma.-collidine
CC1=CC(=NC(=C1)C)C
121.183
121.18
99%

Chemical Identifiers

108-75-8
121.183
BWZVCCNYKMEVEX-UHFFFAOYSA-N
7953
CC1=CC(=NC(=C1)C)C
C8H11N
MFCD00006338
2,4,6-collidine, gamma-collidine, s-collidine, pyridine, 2,4,6-trimethyl, collidine, sym-collidine, 2,4,6-kollidin, unii-7ie4bk5j5v, 2,4,6-trimethyl-pyridine, .gamma.-collidine
2,4,6-trimethylpyridine
Safety and Handling

Safety and Handling

GHS P Statement
Flammable liquid and vapour.
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 203-613-3

RTECSNumber : UU0970000

TSCA : TSCA

Recommended Storage : This product darkens on standing

SDS Documents