missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-[4-(6-Chlorobenzo[d]oxazol-2-yloxy)phenoxy]-N-(2-fluorophenyl)-N-methylpropionamide, 97%

Manufacturer:  Alfa Aesar™ H6351403

 View more versions of this product

Catalog No. AAH6351403


 

Description

Description

Applications
An aryloxyphenoxypropionate (AOPP) post-emergence plant control that shows an exclusive whole plant safety to rice with a high control efficacy to annual grass weeds, especially barnyardgrass.

Solubility
Soluble in methanol, chloroform and dimethyl sulfoxide.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

2-[4-(6-Chlorobenzo[d]oxazol-2-yloxy)phenoxy]-N-(2-fluorophenyl)-N-methylpropionamide
1g
97%
MFCD11111427
ADDQHLREJDZPMT-AWEZNQCLSA-N
(2S)-2-[4-[(6-chloro-1,3-benzoxazol-2-yl)oxy]phenoxy]-N-(2-fluorophenyl)-N-methylpropanamide
59988032
77°C to 81°C
256412-89-2
C23H18ClFN2O4
UN3077
CC(C(=O)N(C)C1=CC=CC=C1F)OC2=CC=C(C=C2)OC3=NC4=C(O3)C=C(C=C4)Cl
440.855
440.86

Chemical Identifiers

256412-89-2
440.855
ADDQHLREJDZPMT-AWEZNQCLSA-N
C23H18ClFN2O4
MFCD11111427
59988032
Safety and Handling

Safety and Handling

GHS H Statement
H332-H317
Harmful if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P304+P340-P312-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

storageNote1 : 2° to 8°C

SDS Documents