Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-[4-(6-Chlorobenzo[d]oxazol-2-yloxy)phenoxy]-N-(2-fluorophenyl)-N-methylpropionamide, 97%

Chemical Identifiers

Chemical Name or Material 2-[4-(6-Chlorobenzo[d]oxazol-2-yloxy)phenoxy]-N-(2-fluorophenyl)-N-methylpropionamide
CAS 256412-89-2
Formula Weight 440.86
MDL Number MFCD11111427
UN Number UN3077
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6351403

  SDS
alfa aesar™
H6351403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6351406

  SDS
alfa aesar™
H6351406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6351414

  SDS
alfa aesar™
H6351414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 440.86
Melting Point 77° to 81°C
Chemical Name or Material 2-[4-(6-Chlorobenzo[d]oxazol-2-yloxy)phenoxy]-N-(2-fluorophenyl)-N-methylpropionamide
Assay 97%
MDL Number MFCD11111427
CAS 256412-89-2
UN Number UN3077
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H332-H317
Harmful if inhaled.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P312-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
DOT Information Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Quantity 1g

Applications
An aryloxyphenoxypropionate (AOPP) post-emergence plant control that shows an exclusive whole plant safety to rice with a high control efficacy to annual grass weeds, especially barnyardgrass.

Solubility
Soluble in methanol, chloroform and dimethyl sulfoxide.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P304+P340-P312-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H332-H317
Harmful if inhaled.
May cause an allergic skin reaction.

2° to 8°C