missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6,8-Tetrachloroquinazoline, 97%

$343.89 - $1,097.99

Chemical Identifiers

CAS 54185-42-1
Molecular Formula C8H2Cl4N2
Molecular Weight (g/mol) 267.918
MDL Number MFCD03844695
InChI Key GYUFSVVBAINEBI-UHFFFAOYSA-N
Synonym quinazoline, 2,4,6,8-tetrachloro
PubChem CID 12703363
IUPAC Name 2,4,6,8-tetrachloroquinazoline
SMILES C1=C(C=C2C(=C1Cl)N=C(N=C2Cl)Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63968MD View Documents Alfa Aesar™
H63968MD
250mg Each for $343.89
 
AAH6396803 View Documents Alfa Aesar™
H6396803
1g Each for $1,097.99
 
Description

Description

Applications
2,4,6,8-Tetrachloroquinazoline is employed as a intermediate for pharmaceutical and organic synthesis.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

54185-42-1
267.918
GYUFSVVBAINEBI-UHFFFAOYSA-N
12703363
C1=C(C=C2C(=C1Cl)N=C(N=C2Cl)Cl)Cl
C8H2Cl4N2
MFCD03844695
quinazoline, 2,4,6,8-tetrachloro
2,4,6,8-tetrachloroquinazoline

Specifications

250mg
C8H2Cl4N2
quinazoline, 2,4,6,8-tetrachloro
C1=C(C=C2C(=C1Cl)N=C(N=C2Cl)Cl)Cl
267.918
267.93
97%
54185-42-1
MFCD03844695
GYUFSVVBAINEBI-UHFFFAOYSA-N
2,4,6,8-tetrachloroquinazoline
12703363
Solid
2,4,6,8-Tetrachloroquinazoline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning