missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4,6,8-Tetrachloroquinazoline, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H63968MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications2,4,6,8-Tetrachloroquinazoline is employed as a intermediate for pharmaceutical and organic synthesis.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
2,4,6,8-Tetrachloroquinazoline | |
C8H2Cl4N2 | |
250 mg | |
GYUFSVVBAINEBI-UHFFFAOYSA-N | |
2,4,6,8-tetrachloroquinazoline | |
12703363 | |
97% |
54185-42-1 | |
MFCD03844695 | |
quinazoline, 2,4,6,8-tetrachloro | |
C1=C(C=C2C(=C1Cl)N=C(N=C2Cl)Cl)Cl | |
267.918 | |
267.93 | |
Solid |
Chemical Identifiers
54185-42-1 | |
267.918 | |
GYUFSVVBAINEBI-UHFFFAOYSA-N | |
12703363 | |
C1=C(C=C2C(=C1Cl)N=C(N=C2Cl)Cl)Cl |
C8H2Cl4N2 | |
MFCD03844695 | |
quinazoline, 2,4,6,8-tetrachloro | |
2,4,6,8-tetrachloroquinazoline |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures