missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,5-Trimethyloxazole, TCI America™

Manufacturer:  TCI America T259525G

 View more versions of this product

Catalog No. T259525G



Specifications

Specifications

2,4,5-Trimethyloxazole
Yellow
20662-84-4
MFCD00005308
2,4,5-trimethyloxazole, trimethyloxazole, oxazole, trimethyl, unii-b04pf51wxi, 2,4,5-trimethyl oxazole, trimethyl-1,3-oxazole, b04pf51wxi, oxazole, 2,4,5-trimethyl, trimethyl-oxazole, pubchem8636
CC1=C(OC(=N1)C)C
111.144
111.14
≥98.0% (GC)
134°C
25g
C6H9NO
1993
ZRLDBDZSLLGDOX-UHFFFAOYSA-N
2,4,5-trimethyl-1,3-oxazole
30215
Clear Liquid at 20°C

Chemical Identifiers

20662-84-4
111.144
ZRLDBDZSLLGDOX-UHFFFAOYSA-N
30215
CC1=C(OC(=N1)C)C
C6H9NO
MFCD00005308
2,4,5-trimethyloxazole, trimethyloxazole, oxazole, trimethyl, unii-b04pf51wxi, 2,4,5-trimethyl oxazole, trimethyl-1,3-oxazole, b04pf51wxi, oxazole, 2,4,5-trimethyl, trimethyl-oxazole, pubchem8636
2,4,5-trimethyl-1,3-oxazole
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3394

TSCA : No

Documents