missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,5-Trimethyloxazole, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1032806

 View more versions of this product

Catalog No. AAA1032806


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4,5-Trimethyloxazole
0.964
33°C (91°F)
C6H9NO
MFCD00005308
UN1993
2,4,5-trimethyloxazole, trimethyloxazole, oxazole, trimethyl, unii-b04pf51wxi, 2,4,5-trimethyl oxazole, trimethyl-1,3-oxazole, b04pf51wxi, oxazole, 2,4,5-trimethyl, trimethyl-oxazole, pubchem8636
CC1=C(OC(=N1)C)C
111.144
111.14
20662-84-4
133°C to 134°C
Pungent
1.442
5 g
107043
ZRLDBDZSLLGDOX-UHFFFAOYSA-N
2,4,5-trimethyl-1,3-oxazole
30215
97%

Chemical Identifiers

20662-84-4
111.144
ZRLDBDZSLLGDOX-UHFFFAOYSA-N
30215
CC1=C(OC(=N1)C)C
C6H9NO
MFCD00005308
2,4,5-trimethyloxazole, trimethyloxazole, oxazole, trimethyl, unii-b04pf51wxi, 2,4,5-trimethyl oxazole, trimethyl-1,3-oxazole, b04pf51wxi, oxazole, 2,4,5-trimethyl, trimethyl-oxazole, pubchem8636
2,4,5-trimethyl-1,3-oxazole
Safety and Handling

Safety and Handling

GHS H Statement
H226-H319-H335
Flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 243-952-4

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only