Please login to your online account to display your discounted pricing

2,4,5-Trimethyloxazole, 95%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  186090250

 View more versions of this product

Catalog No. AC186090250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥93%
Physical Form Liquid
Formula Weight 111.14
Quantity 25g
Color Clear yellow
EINECS Number 243-952-4
TSCA TSCA
Density 0.95g/mL
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 2, 4, 5-Trimethyloxazole
Packaging Glass bottle
Refractive Index 1.4410 to 1.4430 (20°C, 589nm)
Name Note 95%
Assay Percent Range 95%
Molecular Formula C6H9NO
MDL Number MFCD00005308
CAS 20662-84-4
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2, 4, 5-Trimethyloxazole
Name Note 95%
CAS 20662-84-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥93%
Assay Percent Range 95%
Molecular Formula C6H9NO
Formula Weight 111.14
MDL Number MFCD00005308
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.

SDS & Safety Documents