Please login to your online account to display your discounted pricing

2,4,5-Trimethyloxazole, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4, 5-Trimethyloxazole
Name Note 95%
CAS 20662-84-4
CAS Min % 94.0
CAS Max % 100.0
Assay 93% min.
Assay Percent Range 95%
Molecular Formula C6H9NO
Formula Weight 111.14g/mol
MDL Number MFCD00005308
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC186090250

  SDS
thermo fisher scientific
186090250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4410 to 1.4430 (20°C, 589nm)
Density 0.95g/mL
Color Clear yellow
Quantity 25g
Chemical Name or Material 2, 4, 5-Trimethyloxazole
Name Note 95%
CAS 20662-84-4
CAS Min % 94.0
CAS Max % 100.0
Physical Form Clear yellow liquid
Assay 93% min.
Assay Percent Range 95%
Molecular Formula C6H9NO
Formula Weight 111.14g/mol
MDL Number MFCD00005308
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 243-952-4
TSCA TSCA
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.